Learn More
Cayman Chemical ResorufIn benzyl ether 10mg
SDP
Catalog No. 502965793
Encompass_Preferred
A fluorometric probe that acts as a substrate for CYP3A4; typically used near its apparent Km value of 30 µM to screen the inhibition/activation potential of test compounds and to predict potential drug-drug interactions
Catalog No. 50-296-5793 Supplier Cayman Chemical Supplier No. 1807710 MG
A fluorometric probe that acts as a substrate for CYP3A4; typically used near its apparent Km value of 30 µM to screen the inhibition/activation potential of test compounds and to predict potential drug-drug interactions