Learn More
AdipoGen Resorufin butyrate
SDP
Catalog No. 502560123
Encompass_Preferred
Chemical. CAS 15585-42-9. Formula C16H13NO4. MW 283.28. Synthetic. Suitable as fluorogenic substrate for lipases, choline esterases.
Catalog No. 50-256-0123 Supplier AdipoGen Supplier No. CDXR0044M025
Chemical. CAS 15585-42-9. Formula C16H13NO4. MW 283.28. Synthetic. Suitable as fluorogenic substrate for lipases, choline esterases.