Learn More
Medchemexpress LLC Resorufin butyrate 100mg | 15585-42-9 | 283.28 g/mol | C16H13NO4 | 100 MG
SDP
Catalog No. 5000227138
Encompass_Preferred
Resorufin butyrate is a fluorogenic substrate for measuring triglyceride lipase and cholinesterase activity. Enzymatic hydrolysis releases resorufin, a fluorescent reporter (Ex 570 nm, Em 580 nm), enabling sensitive plate-based fluorescence assays. The compound is supplied as a solid with high purity for biochemical and enzymology applications.
  • Fluorogenic substrate for triglyceride lipases and cholinesterase.
  • Excitation 570 nm, emission 580 nm for sensitive fluorescence detection.
  • High purity (99.0%) suitable for biochemical assays.
  • Solid form; recommended storage -20°C sealed; in solvent -80°C for extended stability.
  • Available in a 100 mg pack for laboratory use.
Catalog No. 50-002-27138 Supplier Medchemexpress LLC Supplier No. HY116583100MG
Resorufin butyrate is a fluorogenic substrate for measuring triglyceride lipase and cholinesterase activity. Enzymatic hydrolysis releases resorufin, a fluorescent reporter (Ex 570 nm, Em 580 nm), enabling sensitive plate-based fluorescence assays. The compound is supplied as a solid with high purity for biochemical and enzymology applications.
  • Fluorogenic substrate for triglyceride lipases and cholinesterase.
  • Excitation 570 nm, emission 580 nm for sensitive fluorescence detection.
  • High purity (99.0%) suitable for biochemical assays.
  • Solid form; recommended storage -20°C sealed; in solvent -80°C for extended stability.
  • Available in a 100 mg pack for laboratory use.