Learn More
Medchemexpress LLC Resorufin butyrate 10mg | 15585-42-9 | 283.28 g/mol | C16H13NO4 | 10 MG
SDP
Catalog No. 5000226188
Encompass_Preferred
Resorufin butyrate is a fluorogenic substrate for triglyceride lipases and cholinesterase. Upon enzymatic hydrolysis it releases resorufin, producing a fluorescent signal suitable for microplate and imaging assays (Ex 570 nm, Em 580 nm).
  • Fluorogenic substrate for triglyceride lipases and cholinesterase.
  • Excitation 570 nm and emission 580 nm for fluorescence detection.
  • Releases resorufin upon hydrolysis for sensitive activity assays.
  • Supplied in small-mass vials suitable for biochemical assays.
  • Store sealed at -20°C; in solvent, -80°C for 6 months or -20°C for 1 month.
  • High purity (99.0%) for reliable assay performance.
Catalog No. 50-002-26188 Supplier Medchemexpress LLC Supplier No. HY11658310MG
Resorufin butyrate is a fluorogenic substrate for triglyceride lipases and cholinesterase. Upon enzymatic hydrolysis it releases resorufin, producing a fluorescent signal suitable for microplate and imaging assays (Ex 570 nm, Em 580 nm).
  • Fluorogenic substrate for triglyceride lipases and cholinesterase.
  • Excitation 570 nm and emission 580 nm for fluorescence detection.
  • Releases resorufin upon hydrolysis for sensitive activity assays.
  • Supplied in small-mass vials suitable for biochemical assays.
  • Store sealed at -20°C; in solvent, -80°C for 6 months or -20°C for 1 month.
  • High purity (99.0%) for reliable assay performance.