Learn More
Cayman Chemical Resorufin butyrate, Purity: ≥98%, CAS: 15585-42-9, 10 mg
SDP
Catalog No. 502984183
Encompass_Preferred
A fluorogenic substrate for triglyceride lipases and cholinesterase; resorufin is released upon enzymatic cleavage by triglyceride lipases or cholinesterase and its fluorescence can be used to quantify triglyceride lipase or cholinesterase activity; ex/em = 570/580 nm, respectively
Catalog No. 50-298-4183 Supplier Cayman Chemical Supplier No. 1959210 MG
A fluorogenic substrate for triglyceride lipases and cholinesterase; resorufin is released upon enzymatic cleavage by triglyceride lipases or cholinesterase and its fluorescence can be used to quantify triglyceride lipase or cholinesterase activity; ex/em = 570/580 nm, respectively