Learn More
Medchemexpress LLC Resorufin butyrate 25mg | 15585-42-9 | 25 MG
SDP
Catalog No. 5000308230
Encompass_Preferred
Resorufin butyrate is a fluorogenic substrate used to measure triglyceride lipase and cholinesterase activity. Upon enzymatic hydrolysis the ester is converted to resorufin, which is highly fluorescent (Ex 570 nm, Em 580 nm). Supplied as a 25 mg solid for research use, it is suitable for fluorescence-based enzyme assays and assay development.
  • Fluorogenic substrate for triglyceride lipases and cholinesterase.
  • Generates resorufin detectable at excitation 570 nm and emission 580 nm.
  • High purity (99.0%) supports reproducible assay results.
  • Stable when stored sealed at -20°C; extended stability in solvent at -80°C.
  • Available in multiple package sizes to support assay optimization.
Catalog No. 50-003-08230 Supplier Medchemexpress LLC Supplier No. HY11658325MG
Resorufin butyrate is a fluorogenic substrate used to measure triglyceride lipase and cholinesterase activity. Upon enzymatic hydrolysis the ester is converted to resorufin, which is highly fluorescent (Ex 570 nm, Em 580 nm). Supplied as a 25 mg solid for research use, it is suitable for fluorescence-based enzyme assays and assay development.
  • Fluorogenic substrate for triglyceride lipases and cholinesterase.
  • Generates resorufin detectable at excitation 570 nm and emission 580 nm.
  • High purity (99.0%) supports reproducible assay results.
  • Stable when stored sealed at -20°C; extended stability in solvent at -80°C.
  • Available in multiple package sizes to support assay optimization.