Learn More
Cayman Chemical Resorufin butyrate, Purity: ≥98%, CAS: 15585-42-9, 25 mg
SDP
Catalog No. 502984185
Encompass_Preferred
A fluorogenic substrate for triglyceride lipases and cholinesterase; resorufin is released upon enzymatic cleavage by triglyceride lipases or cholinesterase and its fluorescence can be used to quantify triglyceride lipase or cholinesterase activity; ex/em = 570/580 nm, respectively
Catalog No. 50-298-4185 Supplier Cayman Chemical Supplier No. 1959225 MG
A fluorogenic substrate for triglyceride lipases and cholinesterase; resorufin is released upon enzymatic cleavage by triglyceride lipases or cholinesterase and its fluorescence can be used to quantify triglyceride lipase or cholinesterase activity; ex/em = 570/580 nm, respectively