Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Resorufin butyrate 50mg | 15585-42-9 | 283.28 g/mol | C16H13NO4 | 50 MG
SDP

Catalog No. 5000307996
Encompass_Preferred

Resorufin butyrate is a fluorogenic substrate that releases the fluorescent reporter resorufin upon enzymatic hydrolysis, enabling sensitive measurement of triglyceride lipase and cholinesterase activity (Ex 570 nm, Em 580 nm).

  • Fluorogenic substrate for triglyceride lipases and cholinesterase.
  • Generates resorufin reporter with Ex 570 nm and Em 580 nm.
  • Molecular weight 283.28 g/mol; formula C16H13NO4.
  • Supplied as a solid with reported purity of 99.0%.
  • Recommended storage sealed and protected from moisture; store at -20°C for long-term.
  • Suitable for enzymatic assays and biochemical screening applications.

Catalog No. 50-003-07996 Supplier Medchemexpress LLC Supplier No. HY11658350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Resorufin butyrate is a fluorogenic substrate that releases the fluorescent reporter resorufin upon enzymatic hydrolysis, enabling sensitive measurement of triglyceride lipase and cholinesterase activity (Ex 570 nm, Em 580 nm).

  • Fluorogenic substrate for triglyceride lipases and cholinesterase.
  • Generates resorufin reporter with Ex 570 nm and Em 580 nm.
  • Molecular weight 283.28 g/mol; formula C16H13NO4.
  • Supplied as a solid with reported purity of 99.0%.
  • Recommended storage sealed and protected from moisture; store at -20°C for long-term.
  • Suitable for enzymatic assays and biochemical screening applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.