Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Resorufin butyrate 5mg | 15585-42-9 | 283.28 g/mol | C16H13NO4 | 5 MG
SDP

Catalog No. 5000226147
Encompass_Preferred

Resorufin butyrate is a fluorogenic substrate used to measure triglyceride lipase and cholinesterase activity. Enzymatic hydrolysis releases the fluorescent dye resorufin (Ex 570 nm, Em 580 nm), enabling sensitive fluorescence-based assays and high-throughput screening. The product is supplied at high purity and is accompanied by datasheet and safety documentation.

  • Fluorogenic substrate for triglyceride lipases and cholinesterases.
  • Fluorescence: excitation 570 nm, emission 580 nm.
  • High purity: 99.0%.
  • Available in multiple pack sizes, including 5 MG research packs.
  • Suitable for fluorescence-based enzymatic assays and high-throughput screening.
  • Storage: store sealed at -20°C; in solvent keep at -80°C for up to 6 months or -20°C for up to 1 month.

Catalog No. 50-002-26147 Supplier Medchemexpress LLC Supplier No. HY1165835MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Resorufin butyrate is a fluorogenic substrate used to measure triglyceride lipase and cholinesterase activity. Enzymatic hydrolysis releases the fluorescent dye resorufin (Ex 570 nm, Em 580 nm), enabling sensitive fluorescence-based assays and high-throughput screening. The product is supplied at high purity and is accompanied by datasheet and safety documentation.

  • Fluorogenic substrate for triglyceride lipases and cholinesterases.
  • Fluorescence: excitation 570 nm, emission 580 nm.
  • High purity: 99.0%.
  • Available in multiple pack sizes, including 5 MG research packs.
  • Suitable for fluorescence-based enzymatic assays and high-throughput screening.
  • Storage: store sealed at -20°C; in solvent keep at -80°C for up to 6 months or -20°C for up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.