Learn More
Abcam Resorufin Cellobioside, 5MG
Supplier:  Abcam AB2743075MG
Encompass_Preferred
MW 537.47 g/mol, Purity >95%. Cellulase substrate that releases the highly fluorescent dye resorufin (Ex/Em = 571/585 nm) upon enzyme activity. Resorufin has a low pKa value anabling enzyme measurements can be obtained at low, physiologically relevant, pH values. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-230