Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Resorufin β-D-galactopyranoside | 95079-19-9 | MFCD00064055 | 98.3% | 375.33 | C18H17NO8 | 5 MG
SDP

Catalog No. 5000221919
Encompass_Preferred

Resorufin-β-D-galactopyranoside is a fluorogenic substrate for β-galactosidase that releases resorufin, a red fluorescent reporter, upon enzymatic hydrolysis. It is provided as a high-purity solid suitable for biochemical and cell-based assays, soluble in DMSO, and compatible with standard fluorescence detection methods.

  • Fluorogenic substrate that produces resorufin upon hydrolysis
  • High purity for reliable assay performance
  • Soluble in DMSO; sonication and warming improve solubility
  • Storage-stable when kept dry; solvent-storage recommendations available
  • Suitable for enzyme kinetics, reporter assays, and cell-based assays

Catalog No. 50-002-21919 Supplier Medchemexpress LLC Supplier No. HY1372495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Resorufin-β-D-galactopyranoside is a fluorogenic substrate for β-galactosidase that releases resorufin, a red fluorescent reporter, upon enzymatic hydrolysis. It is provided as a high-purity solid suitable for biochemical and cell-based assays, soluble in DMSO, and compatible with standard fluorescence detection methods.

  • Fluorogenic substrate that produces resorufin upon hydrolysis
  • High purity for reliable assay performance
  • Soluble in DMSO; sonication and warming improve solubility
  • Storage-stable when kept dry; solvent-storage recommendations available
  • Suitable for enzyme kinetics, reporter assays, and cell-based assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.