Learn More
Medchemexpress LLC Resorufin-d6 | 1196157-65-9 | MFCD11656671 | 97.3% | 219.23 | C12HD6NO3 | 1MG
SDP
Catalog No. 5000236426
Encompass_Preferred
Resorufin-d6 is the deuterium-labeled analog of resorufin, a highly fluorescent pink dye used for fluorescence-based detection of reactive oxygen and nitrogen species (ROS/RNS) and related assays. The isotopically labeled compound is intended for use as an internal standard, tracer, or reference in fluorescence and mass spectrometry workflows, providing mass differentiation while retaining resorufin's photophysical properties.
  • Deuterium-labeled for use as an internal standard in analytical assays.
  • Highly fluorescent with photophysical properties matching resorufin.
  • Enables accurate quantification in fluorescence and mass spectrometry workflows.
  • Typical purity reported at about 97.3% per certificate of analysis.
  • Stable when stored sealed at low temperature and protected from moisture and light.
Catalog No. 50-002-36426 Supplier Medchemexpress LLC Supplier No. HY123533S1MG
Resorufin-d6 is the deuterium-labeled analog of resorufin, a highly fluorescent pink dye used for fluorescence-based detection of reactive oxygen and nitrogen species (ROS/RNS) and related assays. The isotopically labeled compound is intended for use as an internal standard, tracer, or reference in fluorescence and mass spectrometry workflows, providing mass differentiation while retaining resorufin's photophysical properties.
  • Deuterium-labeled for use as an internal standard in analytical assays.
  • Highly fluorescent with photophysical properties matching resorufin.
  • Enables accurate quantification in fluorescence and mass spectrometry workflows.
  • Typical purity reported at about 97.3% per certificate of analysis.
  • Stable when stored sealed at low temperature and protected from moisture and light.