Learn More
Cayman Chemical Resorufin β-D-Galactopyranoside, Purity: ≥98%, CAS: 95079-19-9, 10 mg
SDP
Catalog No. 502941309
Encompass_Preferred
A fluorogenic substrate for β-galactosidase; cleaved by β-galactosidase to release the fluorescent moiety resorufin; ex/em = 570/580 nm, respectively
Catalog No. 50-294-1309 Supplier Cayman Chemical Supplier No. 2801510 MG
A fluorogenic substrate for β-galactosidase; cleaved by β-galactosidase to release the fluorescent moiety resorufin; ex/em = 570/580 nm, respectively