Learn More
Cayman Chemical ResorufIn DGalactopyrnosI 25mg
SDP
Catalog No. 502941695
Encompass_Preferred
A fluorogenic substrate for β-galactosidase; cleaved by β-galactosidase to release the fluorescent moiety resorufin; ex/em = 570/580 nm, respectively
Catalog No. 50-294-1695 Supplier Cayman Chemical Supplier No. 2801525 MG
A fluorogenic substrate for β-galactosidase; cleaved by β-galactosidase to release the fluorescent moiety resorufin; ex/em = 570/580 nm, respectively