Learn More
Sigma Aldrich Fine Chemicals Biosciences Resorufin ethyl ether5MG
Catalog No. 502902486
Encompass
Resorufin is the reduced product of resazurin also termed as Alamar Blu. It exhibits fluorescence at longer wavelengths and is flexible for modification in its hydroxyl group.
Catalog No. 50-290-2486 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E37635MG
Resorufin is the reduced product of resazurin also termed as Alamar Blu. It exhibits fluorescence at longer wavelengths and is flexible for modification in its hydroxyl group.