Learn More
Abcam Resorufin ß-D-galactopyranoside, 10MG
Supplier:  Abcam AB27363910MG
Encompass_Preferred
MW 375.3 Da, Purity >95%. Resorufin β-D-galactopyranoside requires only a single-step hydrolysis reaction to attain full fluorescence making it especially useful for sensitive enzyme measurements in applications such as ELISAs or high-content screening systems. Suitable for continuous measurement of enzyme activity at Ex/Em=573/585 nm. Can quantitate beta-galactosidaseactivity in single yeast cells via flow cytometry.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-081