Learn More
Abcam Resorufin maltotriose, 5MG
Supplier:  Abcam AB2753715MG
Encompass_Preferred
MW 669.91 g/mol. Alpha-amylase substrate with red emission (λ max=585nm) after enzyme reaction. Useful for biofuels research.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-512-95