Learn More
Abcam Resorufin oleate, 10MG
Catalog No. 50996221
Encompass_Preferred
MW 493.6 Da, Purity >98%. Fluorogenic lipase substrate. Releases resorufin on hydrolysis. Red fluorescent dye exhibits Ex/Em = 571/585 nm. Resorufin has a low pKa making it suitable for emission readings at low pH values.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-221 Supplier Abcam Supplier No. AB27504410MG
MW 493.6 Da, Purity >98%. Fluorogenic lipase substrate. Releases resorufin on hydrolysis. Red fluorescent dye exhibits Ex/Em = 571/585 nm. Resorufin has a low pKa making it suitable for emission readings at low pH values.
The product is subject to the following: Abcam Restricted Use Statement