Learn More
Medchemexpress LLC Resorufin pentyl ether | 87687-03-4 | 98.2% | 1 MG
SDP
Catalog No. 5000410041
Encompass_Preferred
Resorufin pentyl ether (Pentoxyresorufin) is a Resazurin analogue that functions as a substrate probe to characterize and differentiate between a variety of inducers of cytochromes P-450. It also exhibits bactericidal activity against N. gonorrhoeae.
  • Resazurin analogue
  • Functions as a substrate probe to characterize and differentiate between various inducers of cytochromes P-450
  • Has bactericidal activity against N. gonorrhoeae
  • For research use only
Catalog No. 50-004-10041 Supplier Medchemexpress LLC Supplier No. HYD01471MG
Resorufin pentyl ether (Pentoxyresorufin) is a Resazurin analogue that functions as a substrate probe to characterize and differentiate between a variety of inducers of cytochromes P-450. It also exhibits bactericidal activity against N. gonorrhoeae.
  • Resazurin analogue
  • Functions as a substrate probe to characterize and differentiate between various inducers of cytochromes P-450
  • Has bactericidal activity against N. gonorrhoeae
  • For research use only