Learn More
AdipoGen Resorufin pentyl ether
SDP
Catalog No. 502560142
Encompass_Preferred
Chemical. CAS 87687-03-4. Formula C17H17NO3. MW 283.32. Resorufin pentyl ether 7-Pentoxyresorufin is a fluorogenic substrate used for monitoring cytochrome P450 CYP450 activities in cell extracts and solutions. It is used as substrate for 7-Pentoxyresorufin-O-dealkylase PROD assays. It is used to screen the inhibition/activation potential of test compounds, to predict potential drug-drug interactions, or to monitor other CYP450 activities. Cytochrome P450 oxidase is a large number of evolutionary related oxidative enzymes important in animal, plant, and bacterial physiology. Most cytochromes P450 CYPs have about 500 amino acids and a heme group at the active site. Upon enzymatic cleavage of 7-Pentoxyresorufin by CYP450, the fluorescent hydrolysis product resorufin is released and its fluorescence can be used to quantify enzyme activity. Spectral data of Resorufin lambdaex=571nm, lambdaem=585nm. Resorufin pentyl ether shows also antibacterial properties.
Catalog No. 50-256-0142 Supplier AdipoGen Supplier No. CDXR0015M005
Chemical. CAS 87687-03-4. Formula C17H17NO3. MW 283.32. Resorufin pentyl ether 7-Pentoxyresorufin is a fluorogenic substrate used for monitoring cytochrome P450 CYP450 activities in cell extracts and solutions. It is used as substrate for 7-Pentoxyresorufin-O-dealkylase PROD assays. It is used to screen the inhibition/activation potential of test compounds, to predict potential drug-drug interactions, or to monitor other CYP450 activities. Cytochrome P450 oxidase is a large number of evolutionary related oxidative enzymes important in animal, plant, and bacterial physiology. Most cytochromes P450 CYPs have about 500 amino acids and a heme group at the active site. Upon enzymatic cleavage of 7-Pentoxyresorufin by CYP450, the fluorescent hydrolysis product resorufin is released and its fluorescence can be used to quantify enzyme activity. Spectral data of Resorufin lambdaex=571nm, lambdaem=585nm. Resorufin pentyl ether shows also antibacterial properties.