Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resorufin Pract., Thermo Scientific™

Catalog No. AC418940100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC418940100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC418940100 Supplier Thermo Scientific Chemicals Supplier No. 418940100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-78-9
Molecular Formula C12H7NO3
Molecular Weight (g/mol) 213.19
MDL Number MFCD00128991
InChI Key HSSLDCABUXLXKM-UHFFFAOYSA-N
Synonym resorufin, resorufine, 7-hydroxy-3h-phenoxazin-3-one, 3h-phenoxazin-3-one, 7-hydroxy, 7-hydroxy-3h-phenoxazine-3-one, hydroxyphenazone, us9216974,resorufin, 4-27-00-02263 beilstein handbook reference, resorufin, dye content 95 %
PubChem CID 69462
ChEBI CHEBI:51602
IUPAC Name 7-hydroxy-3H-phenoxazin-3-one
SMILES OC1=CC=C2N=C3C=CC(=O)C=C3OC2=C1

Specifications

Color Black
Infrared Spectrum Authentic
Quantity 10 g
Beilstein 27, 128
Formula Weight 213.19
Physical Form Fine Crystalline Powder
Chemical Name or Material Resorufin, pract.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.