Learn More
Cayman Chemical ResorufIn Stndard 500uL
SDP
Catalog No. 502895169
Encompass_Preferred
A racemic mixture of biotin (R)-sulfoxide and biotin (S)-sulfoxide
Catalog No. 50-289-5169 Supplier Cayman Chemical Supplier No. 700023500 UL
A racemic mixture of biotin (R)-sulfoxide and biotin (S)-sulfoxide