Learn More
Cayman Chemical Resorufin (sodium salt), Purity: ≥98%, CAS: 34994-50-8, 500 mg
SDP
Catalog No. 502988236
Encompass_Preferred
A fluorescent probe; has been used in the synthesis of resorufin-based fluorescence probes; has also been used for the direct measurement of solvent-solute hydrogen bond dynamics with distinct absorption and emission peaks in solvents with strong hydrogen bonds and overlapping peaks in solvents with weak hydrogen bonds; ex/em = 570/580 nm, respectively
Catalog No. 50-298-8236 Supplier Cayman Chemical Supplier No. 30789500 MG
A fluorescent probe; has been used in the synthesis of resorufin-based fluorescence probes; has also been used for the direct measurement of solvent-solute hydrogen bond dynamics with distinct absorption and emission peaks in solvents with strong hydrogen bonds and overlapping peaks in solvents with weak hydrogen bonds; ex/em = 570/580 nm, respectively