Learn More
Sigma Aldrich Fine Chemicals Biosciences ResourceTM S Cytiva 17 118
Catalog No. 502908711
Encompass
RESOURCE(TM) S columns are prepacked with SOURCE(TM) 15S a strong cation exchanger for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.The small (15 mum) monodisperse bead give high-resolution purification at high flow rates. In addition hydrophilization of the beads minimizes nonspecific adsorption and allows high recovery of purified sample. The material of the column body is PEEK (polyetheretherketone). SOURCE(TM) 30Q and 30S media with a 30 mum bead size are designed for intermediate purification and large-scale polishing and allow higher flow rates.
Catalog No. 50-290-8711 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17118001
RESOURCE(TM) S columns are prepacked with SOURCE(TM) 15S a strong cation exchanger for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.The small (15 mum) monodisperse bead give high-resolution purification at high flow rates. In addition hydrophilization of the beads minimizes nonspecific adsorption and allows high recovery of purified sample. The material of the column body is PEEK (polyetheretherketone). SOURCE(TM) 30Q and 30S media with a 30 mum bead size are designed for intermediate purification and large-scale polishing and allow higher flow rates.