Learn More
Grainger Smoke Tube PK6
Catalog No. NC9781934
Encompass_Preferred
Allegro Smoke Tube, Sell Pack: 6 BX, Includes instruction manual, six irritant smoke tubes, aspirator bulb, smoke tube opener and carrying pouch, Conduct test with P100 filters, Confirms 10x fit factor
Catalog No. NC9781934 Supplier Grainger Supplier No. 3NPE4
Allegro Smoke Tube, Sell Pack: 6 BX, Includes instruction manual, six irritant smoke tubes, aspirator bulb, smoke tube opener and carrying pouch, Conduct test with P100 filters, Confirms 10x fit factor