Learn More
Essendant Particulate Respirator 8210V, N95, Cool Flow Valve, Standard Size, 10/Box
Catalog No. 2679590
Encompass_Preferred
Half-face-mask respirator provides excellent protection from non-oil-based particulates. Cool Flow™ exhalation valve reduces heat buildup, keeping the wearer more comfortable. Adjustable nose clip and foam nose cushion help ensure a secure seal and a personalized fit. Electrostatically charged microfiber filter media. Elastic, latex-free two-strap headband. Designed for grinding, sanding, sweeping and other dust-creating tasks.
Catalog No. 26-795-90 Supplier Essendant Supplier No. MMM8210V
Half-face-mask respirator provides excellent protection from non-oil-based particulates. Cool Flow™ exhalation valve reduces heat buildup, keeping the wearer more comfortable. Adjustable nose clip and foam nose cushion help ensure a secure seal and a personalized fit. Electrostatically charged microfiber filter media. Elastic, latex-free two-strap headband. Designed for grinding, sanding, sweeping and other dust-creating tasks.