Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BinaxNOW™ Respiratory Control Swab Kit COVID-19/Flu A and B 10 Positive and 10 Negative Swabs

Catalog No. 23025606 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Format:
Control Swab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
23-025-606 Control Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-025-606 Supplier BinaxNOW™ Supplier No. 20002055
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

COVID-19/Flu Controls (10P/10N)

  • BinaxNOW™ COVID-19/Flu A and B Control Swab Kit is an external qualitative quality control intended for ensuring that reagents and materials are working and that the test procedure is correctly performed.
  • Includes 10 Positive COVID-19/Flu Control Swabs and 10 Negative COVID-19/Flu Control Swabs

Specifications

CE Marker No
Clia Complexity Waived
Detectable Analytes Influenza A, Influenza B, SARS-CoV-2
Detection Method Lateral flow immunoassay
Format Control Swab
Sample Type Anterior Nasal Swab
Type COVID-19/Flu A and B Control Kit
For Use With (Application) Detection and differentiaion of influenza A, influenza B, and SARS-CoV-2
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.