Learn More
ATCC Respiratory Exclusivity Panel
Catalog No. 502383876
Encompass
This panel of seven quantitative genomic and synthetic nucleic acid preparations was selected from respiratory pathogen strains commonly cited in exclusivity or cross-reactivity studies in SARS-CoV-2 EUA submissions and/or FDA 510(k) submissions.
Catalog No. 50-238-3876 Supplier ATCC Supplier No. MP37
This panel of seven quantitative genomic and synthetic nucleic acid preparations was selected from respiratory pathogen strains commonly cited in exclusivity or cross-reactivity studies in SARS-CoV-2 EUA submissions and/or FDA 510(k) submissions.