Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Visby Medical™ Respiratory Health Control Swabs

Catalog No. 23111202 Shop All Visby Medical Products
Change view
Click to view available options
Visby Medical™ Respiratory Control Swabs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-111-202 Visby Medical™ Respiratory Control Swabs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-111-202 Supplier Visby Medical™ Supplier No. PS400381

missing translation for 'validMainframeMsg'

External positive and negative quality controls for use with the Visby Medical™ Respiratory Health Test. Contains 2 positive and 2 negative external control swabs. For use under FDA emergency use authorization only.

  • Intended for use as an external positive and negative quality control to monitor the performance of the Visby Medical™ Respiratory Health Test.
  • 2 positive and 2 negative external control swabs included.
  • For use with the Visby Medical™ Respiratory Health Tests (PS-400380).
  • Refer to Instructions for Use or Contact Visby Customer Experience Team.

Specifications

Format Control Swabs
For Use With (Equipment) Visby Respiratory Health Test, Visby Power Adapter
Sample Type Nasal Swab
Type Control Swab
Quantity 4 Control Swabs
Stability Controlled Room Temperature
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.