Learn More
Visby Medical™ Respiratory Health Starter Kit
Catalog No. 23111200 Shop All Visby Medical Products
Change view
Click to view available options
Visby Medical™ Respiratory Health Kit
Catalog No.
23-111-200 Visby Medical™ Respiratory Health Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-111-200 Supplier Visby Medical™ Supplier No. PS400532
This item is not returnable. View return policy
A bundle to get started with the Visby Medical™ Respiratory Health Test. The starter kit contains, 2 respiratory health test kits (40 tests), 1 power adapter, positive and negative external controls, and training. Swabs purchased separately.
  • Discounted bundle to get started with Visby Medical™ Respiratory Health
  • Includes: 2 Respiratory Health Test Kits (40 tests), 1 Power Adapter, Visby Medical™ Respiratory Health Control Swabs, staff training, and provider-to-provider(s) discussion.
  • Swabs purchased separately.
  • For new customers and first order only.
  • One starter kit per clinic location.
  • Drop-ship only.
Specifications
CE Marker No
Clia Complexity Waived (FDA, EUA)
Sample Type Anterior Nasal or Nasopharyngeal
Type Respiratory Health Starter Kit