Learn More
Visby Medical™ Respiratory Health Test
Catalog No. 23111203 Shop All Visby Medical Products
Change view
Click to view available options
Visby Medical™ Respiratory Health Test
Catalog No.
23-111-203 Visby Medical™ Respiratory Health Test
1 options
This item is not returnable. View return policy
Catalog No. 23-111-203 Supplier Visby Medical™ Supplier No. PS400380
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
RT-PCR point-of care test detecting and differentiating COVID-19, Flu A, and Flu B in under 30 minutes. Test to treat in a single patient visit. For emergency use authorization only.
  • RT-PCR test for COVID-19, Flu A, and Flu B.
  • Instrument-free, portable, and scalable, run as many tests as you have power adapters.
  • Less than 30 minute test time.
  • One swab with three targets.
  • Test to treat in a single patient visit.
  • No capital investment, service contracts or maintenance agreements.
  • Flu A (PPA: 97.9%, NPA: 99.3%); Flu B (PPA: 96.9%, NPA: 100%); COVID (PPA: 97.4%, NPA: 98.8%).
  • NP or AN clinician or self (AN) swab collection.
  • Swabs purchased separately.
  • Room temperature storage.
  • 20 tests per case.
  • Required for use with the Visby Medical™ Power Adapter (PS-000288)
Specifications
CE Marker No
Clia Complexity Waived (FDA)
Type PCR Test
Disposable Single-use