Learn More
Poltex Respiratory Hygiene Station S
SDP
Catalog No. 22000500
Change view
Click to view available options
Mounting Type:
Countertop
Stand
Wall
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
22-000-500 Wall
22-000-498 Countertop
22-000-499 Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Consumables shown for demonstration purposes only and are not included with the product.
Catalog No. 22-000-500 Supplier Poltex Supplier No. RESPSW
Add to Cart
Add to Cart
Consumables shown for demonstration purposes only and are not included with the product.
Poltex™ Locking Respiratory Hygiene Station S holds two boxes of masks (child mask/adult mask or any combination), one large box of tissue or two half-size boxes, one bottle of hand sanitizer, and one 8.5 x 11 in. sign.
  • Keys are all keyed the same
  • Durable 1/4in thick PETG construction.
  • Comes with drip tray
  • See image for dimensions
  • Consumables not included
  • Choose from:
    • Free-standing: Comes with 1/2 in. thick acrylic stand with non-slip rubber bumpons.
    • Wall mount: Sign holder mounts to wall, then station mounts to sign holder.
    • Countertop placement: Comes with 1/2 in. thick acrylic countertop base with non-slip rubber bumpons.
Specifications
Product Type Respiratory Hygiene Station
For Use With (Equipment) General hygiene products
Mounting Type Wall