Learn More
ATCC Respiratory Inclusivity Panel
Catalog No. 502384435
Encompass
This panel of five quantitative genomic RNA and one quantitative, heat-inactivated preparation was selected from viral strains commonly cited in limit-of-detection studies or as control materials in SARS-CoV-2 EUA submissions and/or FDA 510(k) submissions.
Catalog No. 50-238-4435 Supplier ATCC Supplier No. MP36
This panel of five quantitative genomic RNA and one quantitative, heat-inactivated preparation was selected from viral strains commonly cited in limit-of-detection studies or as control materials in SARS-CoV-2 EUA submissions and/or FDA 510(k) submissions.