Learn More
ZeptoMetrix Respiratory Syncytial Virus A Culture Fluid
SDP
Catalog No. 22032239 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-032-239 1 mL
1 options
Catalog No. 22-032-239 Supplier ZeptoMetrix Supplier No. 0810452CFHI
Add to Cart
Add to Cart
Heat inactivated viral culture fluids are sold as consumable testing materials.
  • The suitability and performance characteristics should be determined by your laboratory for each intended usage.
  • These products are not intended for use in the manufacture or processing of injectable products subject to licensure under section 351 of the Public Health Service Act or for any other product intended for administration to humans.
  • For research use only. Not for use in diagnostic procedures.
Specifications
Product Type Virus Control
Sterility Non-sterile
Quantity 1 mL