Learn More
McKesson General Medical Flowflex COVID-19 Antigen Test, Rapid Results in 15 Min, CLIA Waived | 1 Test
Supplier:  McKesson General Medical 1257910
Encompass_Preferred
The Flowflex COVID-19 Antigen Home Test is a rapid test designed for the qualitative detection of the SARS-CoV-2 nucleocapsid protein antigen from nasal swab specimens. This easy-to-use, over-the-counter kit provides results in just 15 minutes, enabling convenient at-home testing for individuals aged 2 and older, with self-collection options for those 14 and older.
Catalog No. NC3779394