Learn More
Waters Corporation Respiratory Viral Panel-SCV2 for BD MAX™System
Catalog No. B445361 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Detectable Analytes:
SARS-CoV-2
Catalog No. Detectable Analytes
B445361 SARS-CoV-2
1 options
Catalog No. B445361 Supplier Waters Corporation Supplier No. 445361
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SARS-CoV-2 viral RNA is generally detectable in NPS and ANS specimens during the acute phase of infection.
  • Automated multiplex real-time RT-PCR test for the qualitative detection of SARS-CoV-2 viral nucleic acid in nasopharyngeal swab (NPS) and anterior nasal swab (ANS) specimens from individuals with signs and symptoms of respiratory tract infection
  • It is intended for use as an aid in the diagnosis of SARS-CoV-2 infection if used in conjunction with other clinical and epidemiological information, and laboratory findings
  • Positive results do not rule out co-infection with other organisms
  • The agent detected by the BD Respiratory Viral Panel-SCV2 for BD MAX™ System may not be the definitive cause of disease
  • Negative results do not preclude SARS-CoV-2 infection
  • The results of this test should not be used as the sole basis for diagnosis, treatment or other patient management decisions
TRUSTED_SUSTAINABILITY
Specifications
CE Marker CE-IVD
Clia Complexity High, Moderate
Detectable Analytes SARS-CoV-2
Type Respiratory Viral Panel
Disposable Single-use
Sterility Sterile
Form Ready-to-Use (RTU)