Learn More
Raine Industries ECG Tab Electrode Resting
Catalog No. NC1804112
Encompass
Comparable to the burdick cardiosens ultra II resting tab electrodes. Silver/silver chloride (Ag/AgCllayer) and solid adhesive gel give patient fast, accurate, clear tracings under all types of conditions. Silver/silver chloride sensing system is defibrillation recoverable. Strong solid adhesive gel with hypoallergenic for consistent tracings. Product and packaging are hypoallergenic. Compatible with all machines. Leaves no residue on patient skin after treatment. Tabs attach to alligator clip connectors at the pin terminations of patient cable lead-wires.
This item is not returnable. View return policy
Catalog No. NC1804112 Supplier Raine Industries Supplier No. PT2334
Comparable to the burdick cardiosens ultra II resting tab electrodes. Silver/silver chloride (Ag/AgCllayer) and solid adhesive gel give patient fast, accurate, clear tracings under all types of conditions. Silver/silver chloride sensing system is defibrillation recoverable. Strong solid adhesive gel with hypoallergenic for consistent tracings. Product and packaging are hypoallergenic. Compatible with all machines. Leaves no residue on patient skin after treatment. Tabs attach to alligator clip connectors at the pin terminations of patient cable lead-wires.