Learn More
Pickering Laboratories Inc Restore™, 250 ml
SDP
Catalog No. NC9652140
Encompass_Preferred
Glyphosate RESTORE™ rapidly removes Iron and all other polyvalent metals from the column and guard. In addition, it preserves the balance of K+/H+ in the resin, thus avoiding a lengthy re-equilibration., 250 ml
Catalog No. NC9652140 Supplier Pickering Laboratories Inc Supplier No. 17000140
Glyphosate RESTORE™ rapidly removes Iron and all other polyvalent metals from the column and guard. In addition, it preserves the balance of K+/H+ in the resin, thus avoiding a lengthy re-equilibration., 250 ml