Learn More
Braintree Scientific Rodent Tail Vein Catheter 1F, 28cm, 23ga needle w/stylet
SDP
Catalog No. NC9278559
Encompass
Restrained Tail Vein Infusion Catheter Easy to insert reduce the risk of tail injury insure intravenous not subcutaneous dosing increase the number of successful tail vein infusions Most researches use butterfly needles to administer fluids via the tail vein. We carry a system that will allow easier access for both timed injections and repeated bolus administration. The MTV system incorporates a small catheter that is easily inserted into the tail vein and will remain insitu in a restrained mouse for as long as restrain is feasible. Due to the flexibility and size of the catheter, trauma to the tail vein is minimized, allowing for long term and repeated bolus dosing. **Sold for research purposes only and not for human use**
Catalog No. NC9278559 Supplier Braintree Scientific Supplier No. MTV1
Restrained Tail Vein Infusion Catheter Easy to insert reduce the risk of tail injury insure intravenous not subcutaneous dosing increase the number of successful tail vein infusions Most researches use butterfly needles to administer fluids via the tail vein. We carry a system that will allow easier access for both timed injections and repeated bolus administration. The MTV system incorporates a small catheter that is easily inserted into the tail vein and will remain insitu in a restrained mouse for as long as restrain is feasible. Due to the flexibility and size of the catheter, trauma to the tail vein is minimized, allowing for long term and repeated bolus dosing. **Sold for research purposes only and not for human use**