Learn More
Med Vet International Jorvet Auspex Avian Restrainer, Medium, J0381B
Catalog No. 5000495476
Encompass_Preferred
Jorvets medium bird restrainer is extremely easy to use. Simply fold the flat restrainer to form a truncated cone shape, and rapidly place over the bird. This bird restrainer is indispensable for various treatments including examinations, administering oral medication, or food, injections, working with legs, feet, or clipping nails.
Catalog No. 50-004-95476 Supplier Med Vet International Supplier No. JOR381B
Jorvets medium bird restrainer is extremely easy to use. Simply fold the flat restrainer to form a truncated cone shape, and rapidly place over the bird. This bird restrainer is indispensable for various treatments including examinations, administering oral medication, or food, injections, working with legs, feet, or clipping nails.