Learn More
Stoelting™ Restraint Cones
SDP
Catalog No. 10000759 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Disposable
Catalog No. Model
10-000-759 Disposable
1 options
Catalog No. 10-000-759 Supplier Stoelting™ Supplier No. 51361
Add to Cart
Add to Cart
Stoelting™ Plastic Restraint Cones hold an animal safely in position for injections through the plastic bag, for decapitation, for cervical dislocation, or for examination of collected field specimens such as field mice or rats. The animal is easily loaded into the cone at the wide end, and heads for the breathing hole at the small end. Sold in packs of 50.
Allows the scientist to avoid contact with feces or urine, and avoids bites and scratches.
Specifications
For Use With (Application) Neuroscience
Material Plastic
Model Disposable
Includes Package of 50