Learn More
Med Vet International Jorvet Nylon Feline Restraint Bag, Large Cat Bag, J0170L
Catalog No. 5000495230
Encompass_Preferred
The Jorvet large feline restraint bags are made of a washable and waterproof nylon material and are designed for proper restraint of a fractious cat. They have a 2-way zipper running full length over the back for front or rear access. This cat restraint bag is equipped with a quick close velcro collar and front leg openings. The pull-tab zipper flaps provide a quick fur-free closure.
Catalog No. 50-004-95230 Supplier Med Vet International Supplier No. JOR170L
The Jorvet large feline restraint bags are made of a washable and waterproof nylon material and are designed for proper restraint of a fractious cat. They have a 2-way zipper running full length over the back for front or rear access. This cat restraint bag is equipped with a quick close velcro collar and front leg openings. The pull-tab zipper flaps provide a quick fur-free closure.