Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Restriction Enzyme Mapping
SDPSCI_ED

Catalog No. S67264 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S67264 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S67264 Supplier Edvotek™ Supplier No. 206
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, Plasmid DNA for Restriction Digest, Restriction Enzyme Reaction Buffer, UltraPure Water, Restriction Enzyme Dilution Buffer, EcoRI Dryzyme, BamHI Dryzyme, DNA Standard Marker, UltraSpec-Agarose™, Electrophoresis Buffer (50X), 10X Gel Loading Solution, InstaStain™ Ethidium Bromide, FlashBlue™ Stain, Microcentrifuge Tubes with attached caps

Requires

  • Requirements:
  • Horizontal gel electrophoresis apparatus
  • Hotplate, micropipetter, balance
  • Waterbath, weigh boats, floating racks
  • White light visualization system
  • Pipets, pipet pump or bulb
  • Glassware, metric rulers, ice
  • All necessary safety equipment
  • Distilled or deionized water

A prerequisite to cloning and DNA sequencing

In this experiment, a plasmid DNA is cleaved with different combinations of restriction enzymes. By determining the fragment size and using agarose gel electrophoresis, the relative positions of the restriction sites can be mapped.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Determining the fragment size and using agarose gel electrophoresis the relative positions of the restriction sites can be mapped
Includes Instructions, hind iii and bgl i restriction enzymes, plasmid dna, buffers, enzyme grade water, standard dna fragments, gel loading solution, agarose, electrophoresis buffer, flashblue™ stain
Quantity 6 sets of restriction digestions
Time Range 1 hour 20 minutes to 1 hour 45 minutes
Content And Storage Refrigerated; Store at: -20°C; -4°F
Class Size 6 Lab Groups
Product Type Mapping Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.