Learn More
Thermo Scientific™ Resuspension Solution for GeneJET Plasmid Maxiprep Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific GeneJET Resuspension Buffer is a component of the GeneJET Plasmid Maxiprep Kit (Cat. No. K0491/K0492) and may be purchased separately.
Specifications
Specifications
| Buffer | Suspension Buffer |
| Format | Kit |
| Product Line | GeneJET |
| Product Type | Resuspension Solution |
| Quantity | 240 mL |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.