Learn More
Apexbio Technology LLC Resveratrol 501-36-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062590
Encompass_Preferred
Resveratrol is a naturally occurring polyphenolic phytoalexin known to function as an activator of the NAD-dependent protein deacetylase SIRT1 Through SIRT1 activation resveratrol modulates downstream pathways implicated in cellular metabolism inflammation aging and tumorigenesis Commonly employed in biomedical research settings resveratrol serves as a molecular tool to explore the regulatory roles and signaling mechanisms involving sirtuin-mediated pathways Cellular assays have demonstrated resveratrol s inhibitory activity against multiple targets with reported IC50 values typically ranging from 20 to 50 M depending on cell type and experimental conditions Therefore resveratrol is widely utilized in vitro and in vivo to elucidate its roles in metabolic regulation inflammation responses cardiovascular function and cancer biology
Catalog No. 50-000-62590 Supplier Apexbio Technology LLC Supplier No. A41825.1
 Restricted Item
Add to Cart
Add to Cart
Resveratrol is a naturally occurring polyphenolic phytoalexin known to function as an activator of the NAD-dependent protein deacetylase SIRT1 Through SIRT1 activation resveratrol modulates downstream pathways implicated in cellular metabolism inflammation aging and tumorigenesis Commonly employed in biomedical research settings resveratrol serves as a molecular tool to explore the regulatory roles and signaling mechanisms involving sirtuin-mediated pathways Cellular assays have demonstrated resveratrol s inhibitory activity against multiple targets with reported IC50 values typically ranging from 20 to 50 M depending on cell type and experimental conditions Therefore resveratrol is widely utilized in vitro and in vivo to elucidate its roles in metabolic regulation inflammation responses cardiovascular function and cancer biology