Learn More
Medchemexpress LLC HY-16561 500mg , Resveratrol trans-Resveratrol CAS:501-36-0 Purity:98%
SDP
Catalog No. 501492344
Encompass_Preferred
Medchemexpress, HY-16561 500mg Resveratrol trans-Resveratrol CAS:501-36-0 Formula:C14H12O3 IC50: 0.8 μM (Adenylyl cyclase), 1 μM (IKKβ), 3.3 and 5 μM (DNA polymerase α and δ) Purity:98% Resveratrol is one of the numerous polyphenolic compounds found in several vegetal sources, has a wide spectrum of targets with IC 50 of 0.8, 1, 3.3 and 5 μM for adenylyl cyclase, IKKβ , DNA polymerase α and δ, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2344 Supplier Medchemexpress LLC Supplier No. HY16561500MG
Medchemexpress, HY-16561 500mg Resveratrol trans-Resveratrol CAS:501-36-0 Formula:C14H12O3 IC50: 0.8 μM (Adenylyl cyclase), 1 μM (IKKβ), 3.3 and 5 μM (DNA polymerase α and δ) Purity:98% Resveratrol is one of the numerous polyphenolic compounds found in several vegetal sources, has a wide spectrum of targets with IC 50 of 0.8, 1, 3.3 and 5 μM for adenylyl cyclase, IKKβ , DNA polymerase α and δ, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.