Learn More
AdipoGen Resveratrol
SDP
Catalog No. 502560119
Encompass_Preferred
Chemical. CAS 501-36-0. Formula C14H12O3. MW 228.24. Synthetic Selective inhibitor of COX−1. Resveratrol is a phenolic phytoalexin found in grape skin and other plants. It has intracellular antioxidant activity and activates SIRT1, a NAD-dependent histone deacetylase involved in mitochondrial biogenesis and the enhancement of peroxisome proliferator-gamma-activated receptor coactivator-1alpha PGC-1alpha and FOXO activity. The anti-diabetic, neuroprotective and anti-adipogenic actions of resveratrol may be mediated via SIRT1 activation.
Catalog No. 50-256-0119 Supplier AdipoGen Supplier No. CDXR0057M100
Chemical. CAS 501-36-0. Formula C14H12O3. MW 228.24. Synthetic Selective inhibitor of COX−1. Resveratrol is a phenolic phytoalexin found in grape skin and other plants. It has intracellular antioxidant activity and activates SIRT1, a NAD-dependent histone deacetylase involved in mitochondrial biogenesis and the enhancement of peroxisome proliferator-gamma-activated receptor coactivator-1alpha PGC-1alpha and FOXO activity. The anti-diabetic, neuroprotective and anti-adipogenic actions of resveratrol may be mediated via SIRT1 activation.