Learn More
AdipoGen Resveratrol
SDP
Supplier:  AdipoGen AGCN20033M500
Encompass_Preferred
Chemical. CAS 501-36-0. Formula C14H12O3. MW 228.2. Isolated from Polygonum cuspidatum. Potent phenolic antioxidant found in grapes and red wine. Eicosanoid synthesis and platelet aggregation inhibitor. Estrogen receptor agonist. Chemopreventive. Specific inhibitor of cyclooxygenase-1 COX-1. Anti-inflammatory. Ribonucleotide reductase and DNA synthesis. Arrests cell cycle at S/G2 phase. Anticancer and antiproliferative compound. Apoptosis inducer. Protein kinase D inhibitor. Does not inhibit PKC. Autophagy inducer. Potent SIRT1 sirtuin 1 activator. Neuroprotective. Adipogenesis inhibitor. PGC-1alpha activator. Sonic hedgehog Shh signaling pathway modulator. Gli1 mRNA expression inhibitor. Downregulates Gli transcriptional activity. Cardioprotective. Anti-diabetic. Senescence modulator. Extends lifespan. Inhibitor of NLRP3 inflammasome activation.
Catalog No. 50-256-2690