Learn More
Abcam Resveratrol, Antio x idant polyphenol, 500MG
Catalog No. 502707331
Encompass_Preferred
MW 228.24 Da, Purity >99%. Polyphenol found in grape skins and red wine. Displays a multitude of biological activities including antioxidant, antiplatelet, antitumor, anti-inflammatory and cardioprotective effects. Additionally, shows antiamyloidogenic and fibril-destabilizing activities.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7331 Supplier Abcam Supplier No. AB120726500MG
MW 228.24 Da, Purity >99%. Polyphenol found in grape skins and red wine. Displays a multitude of biological activities including antioxidant, antiplatelet, antitumor, anti-inflammatory and cardioprotective effects. Additionally, shows antiamyloidogenic and fibril-destabilizing activities.
The product is subject to the following: Abcam Restricted Use Statement