Learn More
Medchemexpress LLC Resveratrol-d4 | 1089051-56-8 | 99.36% | 232.27 | 1 MG
SDP
Catalog No. 5000391781
Encompass_Preferred
Resveratrol-d4 is a deuterium-labeled version of Resveratrol, a natural polyphenolic phytoalexin known for its anti-oxidant, anti-inflammatory, cardioprotective, and anti-cancer properties. It targets various biological pathways including mTOR, JAK, and β-amyloid, and acts as a specific SIRT1 activator. This compound also functions as a potent pregnane X receptor (PXR) inhibitor and an Nrf2 activator, helping to ameliorate aging-related progressive renal injury in mouse models and increasing nitric oxide production in endothelial cells.
  • Deuterium-labeled for tracer applications
  • Suitable as an internal standard for quantitative analysis
  • Exhibits anti-oxidant and anti-inflammatory properties
  • Supports cardioprotective and anti-cancer effects
  • Activates SIRT1 and Nrf2 pathways
Catalog No. 50-003-91781 Supplier Medchemexpress LLC Supplier No. HY16561S1MG
Resveratrol-d4 is a deuterium-labeled version of Resveratrol, a natural polyphenolic phytoalexin known for its anti-oxidant, anti-inflammatory, cardioprotective, and anti-cancer properties. It targets various biological pathways including mTOR, JAK, and β-amyloid, and acts as a specific SIRT1 activator. This compound also functions as a potent pregnane X receptor (PXR) inhibitor and an Nrf2 activator, helping to ameliorate aging-related progressive renal injury in mouse models and increasing nitric oxide production in endothelial cells.
  • Deuterium-labeled for tracer applications
  • Suitable as an internal standard for quantitative analysis
  • Exhibits anti-oxidant and anti-inflammatory properties
  • Supports cardioprotective and anti-cancer effects
  • Activates SIRT1 and Nrf2 pathways